accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 723753-84-2

HNClF

CAS RN 723753-84-2

IUPAC Name: 4-chloro-N-[(2-fluorophenyl)methyl]aniline
Synonyms: 4-chloro-N-[(2-fluorophenyl)methyl]aniline
Molecular Weight (g/mol): 235.69
Molecular Formula: C13H11ClFN
InChi Key: QCLJXZCWBRKJHD-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1CNC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results