accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7223-50-9

ONHCO

CAS RN 7223-50-9

IUPAC Name: 2-(prop-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(prop-2-yn-1-yl)isoindole-1,3-dione
Molecular Weight (g/mol): 185.18
Molecular Formula: C11H7NO2
InChi Key: PAZCLCHJOWLTGA-UHFFFAOYSA-N
SMILES: O=C1N(CC#C)C(=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results