accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

POHOOHNNN

CAS RN 721457-32-5

IUPAC Name: (12-azidododecyl)phosphonic acid
Synonyms: 12-azidododecylphosphonic acid
Molecular Weight (g/mol): 291.33
Molecular Formula: C12H26N3O3P
InChi Key: WHRLXOVKIALVOF-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCCCCCCCCCCCN=[N+]=[N-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results