CAS RN 72104-34-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N2-Methyl Alfuzosin, TRC
CAS: 72104-34-8 Molecular Formula: C19 H27 N5 O4 . Cl H Molecular Weight (g/mol): 425.91 Synonym: Alfuzosin Hydrochloride Imp. C (EP) as Hydrochloride,(2RS)-N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)amino]propyl]-N-methyltetrahydrofuran-2-carboxamide Hydrochloride IUPAC Name: N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)amino]propyl]-N-methyloxolane-2-carboxamide;hydrochloride SMILES: Cl.COc1cc2nc(NCCCN(C)C(=O)C3CCCO3)nc(N)c2cc1OC