accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 71985-80-3

CH3NOHOHCl

CAS RN 71985-80-3

IUPAC Name: 1-methylpiperidine-4-carboxylic acid hydrochloride
Synonyms: 1-methylpiperidine-4-carboxylic acid hydrochloride
Molecular Weight (g/mol): 179.64
Molecular Formula: C7H14ClNO2
InChi Key: NLUDEWJJEMHIIL-UHFFFAOYSA-N
SMILES: Cl.CN1CCC(CC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results