accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOFN

CAS RN 719-70-0

IUPAC Name: 1-(4-fluoro-2-nitrophenyl)piperidine
Synonyms: 1-(4-fluoro-2-nitrophenyl)piperidine
Molecular Weight (g/mol): 224.24
Molecular Formula: C11H13FN2O2
InChi Key: YSBDMNNCMGUEAL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CC=C1N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results