accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONSONNOFFOCH3Na

CAS RN 718635-09-7

IUPAC Name: sodium 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methanesulfinyl]-1H-1,3-benzodiazol-1-ide hydrate
Synonyms: sodium pantoprazole(1-) hydrate
Molecular Weight (g/mol): 423.37
Molecular Formula: C16H16F2N3NaO5S
InChi Key: CGJRLPRCWSHOFU-UHFFFAOYNA-N
SMILES: O.[Na+].COC1=C(OC)C(CS(=O)C2=NC3=CC(OC(F)F)=CC=C3[N-]2)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results