accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNO

CAS RN 7179-93-3

IUPAC Name: methyl[(oxolan-3-yl)methyl]amine
Synonyms: methyl(oxolan-3-ylmethyl)amine
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: ZUGRRINNTXSWBR-UHFFFAOYNA-N
SMILES: CNCC1CCOC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results