accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOOHOOOCH3OHOH2O

CAS RN 71607-31-3

IUPAC Name: 2,3-bis(4-methylbenzoyloxy)butanedioic acid hydrate
Synonyms: 2,3-bis(4-methylbenzoyloxy)butanedioic acid hydrate
Molecular Weight (g/mol): 404.37
Molecular Formula: C20H20O9
InChi Key: FOTRUJUPLHRVNU-UHFFFAOYNA-N
SMILES: O.CC1=CC=C(C=C1)C(=O)OC(C(OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results