accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CHC

CAS RN 7154-75-8

IUPAC Name: 4-methylpent-1-yne
Synonyms: 1-pentyne, 4-methyl-
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: OXRWICUICBZVAE-UHFFFAOYSA-N
SMILES: CC(C)CC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)