accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7149-49-7

OO

CAS RN 7149-49-7

IUPAC Name: naphthalene-2,3-dicarbaldehyde
Synonyms: naphthalene-2,3-dicarbaldehyde
Molecular Weight (g/mol): 184.19
Molecular Formula: C12H8O2
InChi Key: ZIPLKLQPLOWLTM-UHFFFAOYSA-N
SMILES: O=CC1=C(C=O)C=C2C=CC=CC2=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)