accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COO

CAS RN 713-95-1

IUPAC Name: 6-heptyloxan-2-one
Synonyms: delta-dodecalactone
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: QRPLZGZHJABGRS-UHFFFAOYNA-N
SMILES: CCCCCCCC1CCCC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results