CAS RN 71239-85-5
CAS RN 71239-85-5
IUPAC Name:
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoate
Synonyms:
(2S)-2-[(tert-butoxycarbonyl)amino]-3-(pyridin-2-yl)propanoate
Molecular Weight (g/mol):
265.29
Molecular Formula:
C13H17N2O4
InChi Key:
KMODKKCXWFNEIK-JTQLQIEISA-M
SMILES:
CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=N1)C([O-])=O
1
–
1
of
1
results
1
N-Boc-3-(2-pyridyl)-L-alanine, 98%
CAS: 71239-85-5 Molecular Formula: C13H18N2O4 Molecular Weight (g/mol): 266.297 MDL Number: MFCD00191190 InChI Key: KMODKKCXWFNEIK-JTQLQIEISA-N Synonym: boc-l-2-pyridylalanine,s-2-tert-butoxycarbonyl amino-3-pyridin-2-yl propanoic acid,boc-3-2-pyridyl-l-alanine,boc-l-3-2-pyridyl-alanine,boc-ala 2-pyridyl-oh,boc-2-pal-oh,boc-3-2-pyridyl-ala-oh,boc-2'-pyridyl-l-ala,n-boc-3-2-pyridyl-l-alanine,boc-l-2-pyridylala PubChem CID: 2734482 IUPAC Name: (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-2-ylpropanoic acid SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=N1)C(=O)O