accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7111-68-4

CH3O

CAS RN 7111-68-4

IUPAC Name: 2'-methyl-[1,1'-biphenyl]-2-carbaldehyde
Synonyms: 2'-methyl-[1,1'-biphenyl]-2-carbaldehyde
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: SEEYJCMPEVOYTF-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results