accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFONHBr

CAS RN 710310-54-6

IUPAC Name: N-(3-bromophenyl)-3-(trifluoromethyl)benzamide
Synonyms: N-(3-bromophenyl)-3-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 344.13
Molecular Formula: C14H9BrF3NO
InChi Key: NUCGNVOUNFOXOQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)C(=O)NC1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results