CAS RN 70978-54-0
IUPAC Name:
1-(5-bromo-2-hydroxy-3-nitrophenyl)ethan-1-one
Synonyms:
1-(5-bromo-2-hydroxy-3-nitrophenyl)ethanone
Molecular Weight (g/mol):
260.04
Molecular Formula:
C8H6BrNO4
InChi Key:
CLNIBJASCGZXHH-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(Br)=CC(=C1O)[N+]([O-])=O
1
–
1
of
1
results
1
5'-Bromo-2'-hydroxy-3'-nitroacetophenone, 97%
CAS: 70978-54-0 Molecular Formula: C8H6BrNO4 Molecular Weight (g/mol): 260.04 MDL Number: MFCD01631134 InChI Key: CLNIBJASCGZXHH-UHFFFAOYSA-N PubChem CID: 689071 IUPAC Name: 1-(5-bromo-2-hydroxy-3-nitrophenyl)ethan-1-one SMILES: CC(=O)C1=CC(Br)=CC(=C1O)[N+]([O-])=O