accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3OHO

CAS RN 7084-93-7

IUPAC Name: 4-(propan-2-yl)cyclohexane-1-carboxylic acid
Synonyms: 4-isopropylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: YRQKWRUZZCBSIG-UHFFFAOYSA-N
SMILES: CC(C)C1CCC(CC1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results