accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNNBr

CAS RN 705263-10-1

IUPAC Name: 6-bromopyrazolo[1,5-a]pyrimidine
Synonyms: 6-bromopyrazolo[1,5-a]pyrimidine
Molecular Weight (g/mol): 198.02
Molecular Formula: C6H4BrN3
InChi Key: VDHTXLUCUNPVLO-UHFFFAOYSA-N
SMILES: BrC1=CN2N=CC=C2N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results