accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 704-91-6

OOHHNN

CAS RN 704-91-6

IUPAC Name: 1H-indazole-6-carboxylic acid
Synonyms: 1H-indazole-6-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: DNCVTVVLMRHJCJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=NNC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results