accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 70102-34-0

IUPAC Name: 3-nitrobenzene-1-carbothioamide
Synonyms: 3-nitrobenzenecarbothioamide
Molecular Weight (g/mol): 182.2
Molecular Formula: C7H6N2O2S
InChi Key: HDQCHDWHHGEXQE-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results