accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOFBr

CAS RN 701-45-1

IUPAC Name: 2-bromo-1-fluoro-4-nitrobenzene
Synonyms: 2-bromo-1-fluoro-4-nitrobenzene
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: FAWMTDSAMOCUAR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results