accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 70091-75-7

H3COOONOO

CAS RN 70091-75-7

IUPAC Name: ethyl 2-(4-nitrophenyl)-2-oxoacetate
Synonyms: ethyl 2-(4-nitrophenyl)-2-oxoacetate
Molecular Weight (g/mol): 223.18
Molecular Formula: C10H9NO5
InChi Key: ZFCXKZCKJZFZGR-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C1=CC=C(C=C1)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results