accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 699012-88-9

OOOOOOCHHCCHScH3CH3CCH3H3CCH3H3C

CAS RN 699012-88-9

IUPAC Name: scandium(3+) tris(4-oxopent-2-en-2-olate) hydrate
Synonyms: scandium(3+) tris(4-oxopent-2-en-2-olate) hydrate
Molecular Weight (g/mol): 360.3
Molecular Formula: C15H23O7Sc
InChi Key: LDRQBGBMJVMLGE-UHFFFAOYSA-K
SMILES: O.[Sc+3].CC([O-])=CC(C)=O.CC([O-])=CC(C)=O.CC([O-])=CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results