CAS RN 699-97-8
CAS RN 699-97-8
IUPAC Name:
[3-(hydroxymethyl)bicyclo[2.2.1]hept-5-en-2-yl]methanol
Synonyms:
[3-(hydroxymethyl)bicyclo[2.2.1]hept-5-en-2-yl]methanol
Molecular Weight (g/mol):
154.21
Molecular Formula:
C9H14O2
InChi Key:
IGHHPVIMEQGKNE-UHFFFAOYNA-N
SMILES:
OCC1C2CC(C=C2)C1CO
1
–
1
of
1
results
1
5-Norbornene-2-endo,3-endo-dimethanol, 98%
CAS: 699-97-8 Molecular Formula: C9H14O2 Molecular Weight (g/mol): 154.209 MDL Number: MFCD00674044 InChI Key: IGHHPVIMEQGKNE-WZENYGAOSA-N Synonym: 5-norbornene-2-endo,3-endo-dimethanol,norborna-5-ene-2alpha,3alpha-dimethanol,bicyclo 2.2.1 hept-5-ene-2,3-dimethanol, 2r,3s-rel-9ci,2s,3r-3-hydroxymethyl bicyclo 2.2.1 hept-5-en-2-yl methanol PubChem CID: 12253651 IUPAC Name: [(2R,3S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol SMILES: C1C2C=CC1C(C2CO)CO