accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 699-08-1

CH3OI

CAS RN 699-08-1

IUPAC Name: 1-ethoxy-4-iodobenzene
Synonyms: 4-iodophenetole
Molecular Weight (g/mol): 248.06
Molecular Formula: C8H9IO
InChi Key: VSIIHWOJPSSIDI-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(I)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results