accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 69847-45-6

(S)ONH2OONaNa

CAS RN 69847-45-6

IUPAC Name: disodium 2-amino-3-(4-oxidophenyl)propanoate
Synonyms: disodium tyrosinate
Molecular Weight (g/mol): 225.16
Molecular Formula: C9H9NNa2O3
InChi Key: ASIYFCYUCMQNGK-UHFFFAOYNA-L
SMILES: [Na+].[Na+].NC(CC1=CC=C([O-])C=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)