CAS RN 69839-83-4
CAS RN 69839-83-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Didox, MedChemExpress
MedChemExpress Didox (NSC-324360) is a synthetic ribonucleotide reductase (RR) inhibitor.