accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6973-60-0

CH3NHOO

CAS RN 6973-60-0

IUPAC Name: 1-methyl-1H-pyrrole-2-carboxylic acid
Synonyms: 1-methylpyrrole-2-carboxylic acid
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: ILAOVOOZLVGAJF-UHFFFAOYSA-N
SMILES: CN1C=CC=C1C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results