accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2ClBrCl

CAS RN 697-88-1

IUPAC Name: 4-bromo-2,6-dichloroaniline
Synonyms: 4-bromo-2,6-dichloroaniline
Molecular Weight (g/mol): 240.91
Molecular Formula: C6H4BrCl2N
InChi Key: NPQBZKNXJZARBJ-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(Br)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results