accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOS

CAS RN 6964-21-2

IUPAC Name: 2-(thiophen-3-yl)acetic acid
Synonyms: 3-thiopheneacetic acid 3-thiophene acetic acid
Molecular Weight (g/mol): 142.17
Molecular Formula: C6H6O2S
InChi Key: RCNOGGGBSSVMAS-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CSC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)