accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COCl

CAS RN 69614-95-5

IUPAC Name: 1-[4-(2-chloroethyl)phenyl]ethan-1-one
Synonyms: 1-[4-(2-chloroethyl)phenyl]ethanone
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: RGVUACHOYCYNMB-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(CCCl)C=C1
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results