accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 69584-87-8

FFN

CAS RN 69584-87-8

IUPAC Name: 2-(2,5-difluorophenyl)acetonitrile
Synonyms: 2-(2,5-difluorophenyl)acetonitrile
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H5F2N
InChi Key: UIMMFRUOZOWROM-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(CC#N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results