accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COON

CAS RN 69555-14-2

IUPAC Name: ethyl 2-[(diphenylmethylidene)amino]acetate
Synonyms: ethyl 2-[(diphenylmethylidene)amino]acetate
Molecular Weight (g/mol): 267.33
Molecular Formula: C17H17NO2
InChi Key: QUGJYNGNUBHTNS-UHFFFAOYSA-N
SMILES: CCOC(=O)CN=C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)