accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 69500-64-7

NHH2N

CAS RN 69500-64-7

IUPAC Name: 1-(pyrrolidin-2-yl)methanamine
Synonyms: 2-pyrrolidinemethanamine
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: AUKXFNABVHIUAC-UHFFFAOYNA-N
SMILES: NCC1CCCN1
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results