accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2NN

CAS RN 69491-59-4

IUPAC Name: 3-(pyrimidin-5-yl)aniline
Synonyms: 3-(pyrimidin-5-yl)aniline
Molecular Weight (g/mol): 171.2
Molecular Formula: C10H9N3
InChi Key: DZEIKJMNXHOFHL-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C1=CN=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results