accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6943-65-3

IUPAC Name: {[2-(carbamimidoylsulfanyl)ethyl]sulfanyl}methanimidamide hydrobromidyl
Synonyms: [2-(carbamimidoylsulfanyl)ethyl]sulfanylmethanimidamide hydrobromidyl
Molecular Weight (g/mol): 258.18
Molecular Formula: C4H10BrN4S2
InChi Key: NPUQATSZCVUYHC-UHFFFAOYSA-N
SMILES: [Br].NC(=N)SCCSC(N)=N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results