accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 69373-37-1

IUPAC Name: 7-nitro-3,4-dihydro-2H-1,4-benzothiazine
Synonyms: 7-nitro-3,4-dihydro-2H-1,4-benzothiazine
Molecular Weight (g/mol): 196.22
Molecular Formula: C8H8N2O2S
InChi Key: LCOAYUPAESYAHF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C(NCCS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results