accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6921-29-5

CHNCHCH

CAS RN 6921-29-5

IUPAC Name: tris(prop-2-yn-1-yl)amine
Synonyms: tripropynyl-amine
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: ZHOBJWVNWMQMLF-UHFFFAOYSA-N
SMILES: C#CCN(CC#C)CC#C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)