accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 690634-81-2

NHONHCl

CAS RN 690634-81-2

IUPAC Name: 2-(pyrrolidine-1-carbonyl)piperidine hydrochloride
Synonyms: 2-(pyrrolidine-1-carbonyl)piperidine hydrochloride
Molecular Weight (g/mol): 218.73
Molecular Formula: C10H19ClN2O
InChi Key: RIAUQORBEIQABE-UHFFFAOYNA-N
SMILES: Cl.O=C(C1CCCCN1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results