accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 690632-37-2

CH3NCH3NOOCH3

CAS RN 690632-37-2

IUPAC Name: methyl 6-(diethylamino)pyridine-3-carboxylate
Synonyms: methyl 6-(diethylamino)pyridine-3-carboxylate
Molecular Weight (g/mol): 208.26
Molecular Formula: C11H16N2O2
InChi Key: MYJCIGZNMMQDRA-UHFFFAOYSA-N
SMILES: CCN(CC)C1=NC=C(C=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results