accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 690632-33-8

CH3HNH2NNO(E/Z)

CAS RN 690632-33-8

IUPAC Name: 1-(3-methyl-1,2-dihydropyridin-2-ylidene)-1-nitrosomethanamine
Synonyms: 1-(3-methyl-1H-pyridin-2-ylidene)-1-nitrosomethanamine
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: YIBWTWSINGSDFJ-UHFFFAOYSA-N
SMILES: CC1=CC=CNC1=C(N)N=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results