accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOH3CCH3H3CH3CNSCH3

CAS RN 690632-24-7

IUPAC Name: 4-methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
Synonyms: 4-methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 301.21
Molecular Formula: C16H20BNO2S
InChi Key: QJROOQWDMKQWEZ-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC=CC=C1)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results