accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOO

CAS RN 69026-14-8

IUPAC Name: 3-(benzyloxy)benzoate
Synonyms: 3-(benzyloxy)benzoate
Molecular Weight (g/mol): 227.24
Molecular Formula: C14H11O3
InChi Key: CISXCTKEQYOZAM-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(OCC2=CC=CC=C2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results