accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 69-74-9

NH2NON(R)O(R)HO(S)HO(S)OHHCl

CAS RN 69-74-9

IUPAC Name: hydrogen 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one chloride
Synonyms: hydrogen cytarabine chloride
Molecular Weight (g/mol): 279.68
Molecular Formula: C9H14ClN3O5
InChi Key: KCURWTAZOZXKSJ-JBMRGDGGSA-N
SMILES: [H+].[Cl-].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
Physical Form 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)