accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 68988-92-1

CH3NCH3NNH3CCH3SOBrHOBrBrOHBrOOH(?)

CAS RN 68988-92-1

IUPAC Name: 2,4,5,7-tetrabromo-9-(2-carboxyphenyl)-3,6-dihydroxy-10λ⁴-xanthen-10-ylium; 7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium
Synonyms: 2,4,5,7-tetrabromo-9-(2-carboxyphenyl)-3,6-dihydroxy-10λ⁴-xanthen-10-ylium; 7-(dimethylamino)-N,N-dimethylphenothiazin-3-iminium
Molecular Weight (g/mol): 933.3
Molecular Formula: C36H27Br4N3O5S
InChi Key: AXIKDPDWFVPGOD-UHFFFAOYSA-O
SMILES: CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C.OC(=O)C1=CC=CC=C1C1=C2C=C(Br)C(O)=C(Br)C2=[O+]C2=C(Br)C(O)=C(Br)C=C12
Quantity 
  • (1)
  • (2)
  • (2)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)