accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 68938-92-1

BrPtBrBrBr

CAS RN 68938-92-1

IUPAC Name: platinum(4+) tetrabromide
Synonyms: platinum(IV) bromide
Molecular Weight (g/mol): 514.7
Molecular Formula: Br4Pt
InChi Key: SNPHNDVOPWUNON-UHFFFAOYSA-J
SMILES: [Br-].[Br-].[Br-].[Br-].[Pt+4]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results