CAS RN 68856-96-2
CAS RN 68856-96-2
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}-3-(4-methoxyphenyl)propanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]-3-(4-methoxyphenyl)propanoic acid
Molecular Weight (g/mol):
295.34
Molecular Formula:
C15H21NO5
InChi Key:
SLWWWZWJISHVOU-UHFFFAOYNA-N
SMILES:
COC1=CC=C(CC(NC(=O)OC(C)(C)C)C(O)=O)C=C1
1
–
1
of
1
results
1
N-Boc-4-methoxy-D-phenylalanine, 95%
CAS: 68856-96-2 Molecular Formula: C15H21NO5 Molecular Weight (g/mol): 295.335 InChI Key: SLWWWZWJISHVOU-GFCCVEGCSA-N Synonym: boc-d-tyr me-oh,boc-4-methoxy-d-phenylalanine,boc-o-methyl-d-tyrosine,boc-4-methoxy-d-phe-oh,n-boc-4-methoxy-d-phenylalanine,2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl propanoic acid,d-tyrosine,n-1,1-dimethylethoxy carbonyl-o-methyl,d-tyrosine, n-1,1-dimethylethoxy carbonyl-o-methyl,boc-l-phe 4-och3-oh,boc-d-phe 4-ome-oh PubChem CID: 7017908 IUPAC Name: (2R)-3-(4-methoxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid SMILES: CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)OC)C(=O)O