accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OBOCH3OCH3

CAS RN 688-71-1

IUPAC Name: tripropyl borate
Synonyms: boric acid (H3BO3), tripropyl ester
Molecular Weight (g/mol): 188.07
Molecular Formula: C9H21BO3
InChi Key: LTEHWCSSIHAVOQ-UHFFFAOYSA-N
SMILES: CCCOB(OCCC)OCCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)