accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 68781-13-5

OOHNH2HCl

CAS RN 68781-13-5

IUPAC Name: 1-aminocyclopropane-1-carboxylic acid hydrochloride
Synonyms: 1-aminocyclopropane-1-carboxylic acid hydrochloride
Molecular Weight (g/mol): 137.56
Molecular Formula: C4H8ClNO2
InChi Key: WTZRHZRMLYNBRM-UHFFFAOYSA-N
SMILES: Cl.NC1(CC1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)