accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CO

CAS RN 6853-57-2

IUPAC Name: 4-pentylbenzaldehyde
Synonyms: 4-pentylbenzaldehyde
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: NQVZPRUSNWNSQH-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results